ENA341294613 Enamine

2-(4-chlorophenyl)-2-methoxyethan-1-amine hydrochloride

  • MDL number MFCD20501938
  • Cas Number : 1354954-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90537

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90537
Availability details for
Quantity
  • Availability
  • Add to Cart