ENA341294618 Enamine

5,6-dimethyl-2,3-dihydro-1-benzofuran-2-carboxylic acid

  • MDL number MFCD20501943
  • Cas Number : 1354950-79-0
  • Molecular Weight : 192.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91284
Availability details for
Quantity
  • Availability
  • Add to Cart