ENA341294619 Enamine

2-[1-(4-fluorophenyl)-1H-pyrazol-3-yl]ethan-1-amine

  • MDL number MFCD20501944
  • Cas Number : 1354957-77-9
  • Molecular Weight : 205.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91291
Availability details for
Quantity
  • Availability
  • Add to Cart