ENA341294620 Enamine

3-aminooxolane-3-carbonitrile hydrochloride

  • MDL number MFCD20501945
  • Cas Number : 1354963-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91333

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91333
Availability details for
Quantity
  • Availability
  • Add to Cart