ENA341294621 Enamine

4-(chloromethyl)-3,5-dimethyl-1-phenyl-1H-pyrazole hydrochloride

  • MDL number MFCD20501946
  • Cas Number : 1354963-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91431

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91431
Availability details for
Quantity
  • Availability
  • Add to Cart