ENA341294622 Enamine

2-methyl-4H-chromene-3-carboxylic acid

  • MDL number MFCD20501947
  • Cas Number : 1354953-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91445
Availability details for
Quantity
  • Availability
  • Add to Cart