ENA341294623 Enamine

3,3-bis(methylamino)-1-(2-methylphenyl)prop-2-en-1-one

  • MDL number MFCD20501948
  • Cas Number : 1354963-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-91488

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91488
Availability details for
Quantity
  • Availability
  • Add to Cart