ENA341294625 Enamine

2-(2,3-dimethylphenyl)piperidine hydrochloride

  • MDL number MFCD20501950
  • Cas Number : 1354952-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 306...308
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91538

Safety Information

msds https://www.enaminestore.com/catalog/EN300-91538
Availability details for
Quantity
  • Availability
  • Add to Cart