ENA341294631 Enamine

3-[(3-bromophenyl)methyl]pyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD20501957
  • Cas Number : 1354963-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91683
Availability details for
Quantity
  • Availability
  • Add to Cart