ENA341294653 Enamine

3-bromo-1-methyl-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD20501985
  • Cas Number : 65060-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-89
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92022
Availability details for
Quantity
  • Availability
  • Add to Cart