ENA341294655 Enamine

5-(chloromethyl)-3-(1H-1,2,4-triazol-3-yl)-1,2,4-oxadiazole

  • MDL number MFCD09728374
  • Cas Number : 953751-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92031

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92031
Availability details for
Quantity
  • Availability
  • Add to Cart