ENA341294656 Enamine

2-(1-ethoxyethyl)-4-(piperazin-1-ylmethyl)-1,3-thiazole

  • MDL number MFCD20501988
  • Cas Number : 1354957-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92049

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92049
Availability details for
Quantity
  • Availability
  • Add to Cart