ENA341294657 Enamine

sodium 4-hydroxyundecanoate

  • MDL number MFCD20501990
  • Cas Number : 57681-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92087
Availability details for
Quantity
  • Availability
  • Add to Cart