ENA341294659 Enamine

benzyl 2-carbamothioylmorpholine-4-carboxylate

  • MDL number MFCD20501992
  • Cas Number : 1354959-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92123

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92123
Availability details for
Quantity
  • Availability
  • Add to Cart