ENA341294660 Enamine

1-[3-fluoro-4-(4-methyl-1H-pyrazol-1-yl)phenyl]ethan-1-amine hydrochloride

  • MDL number MFCD20501994
  • Cas Number : 1354950-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92133

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92133
Availability details for
Quantity
  • Availability
  • Add to Cart