ENA341294661 Enamine

methyl (3R)-3-amino-3-[3-(trifluoromethyl)phenyl]propanoate hydrochloride

  • MDL number MFCD20501995
  • Cas Number : 1354970-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92137
Availability details for
Quantity
  • Availability
  • Add to Cart