ENA341294665 Enamine

3,3-bis(methylamino)-1-(pyridin-3-yl)prop-2-en-1-one

  • MDL number MFCD20501999
  • Cas Number : 1354951-40-8
  • Molecular Weight : 191.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-92194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92194
Availability details for
Quantity
  • Availability
  • Add to Cart