ENA341294677 Enamine

2-(2-chlorophenyl)acetyl chloride

  • MDL number MFCD00130223
  • Cas Number : 51512-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90191

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-90191
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart