ENA341294692 Enamine

(1-methyl-1H-pyrazol-4-yl)methanesulfonamide

  • MDL number MFCD16679030
  • Cas Number : 1247841-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90207

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90207
Availability details for
Quantity
  • Availability
  • Add to Cart