ENA341294696 Enamine

3-fluoronaphthalene-1-carboxylic acid

  • MDL number MFCD17977213
  • Cas Number : 575-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-187
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90211
Availability details for
Quantity
  • Availability
  • Add to Cart