ENA341294721 Enamine

2-{1-[(4-bromo-3-fluorophenyl)carbonyl]pyrrolidin-2-yl}acetic acid

  • MDL number MFCD16509108
  • Cas Number : 1283631-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90276

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90276
Availability details for
Quantity
  • Availability
  • Add to Cart