ENA341294734 Enamine

1-(4-methyl-2-oxo-2,3-dihydro-1,3-thiazole-5-sulfonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD11550607

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247...249
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90299

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90299
Availability details for
Quantity
  • Availability
  • Add to Cart