ENA341294736 Enamine

3-(1-methyl-1H-1,2,3,4-tetrazol-5-yl)benzene-1-sulfonyl chloride

  • MDL number MFCD11650663
  • Cas Number : 1099660-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87-89
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90301

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
hcodes C
MSDS https://www.enaminestore.com/catalog/EN300-90301
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart