ENA341294739 Enamine

1-[(6-methoxypyridin-3-yl)methyl]-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD17416610
  • Cas Number : 1354951-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90304
Availability details for
Quantity
  • Availability
  • Add to Cart