ENA341294740 Enamine

2-amino-3-(4-bromo-1H-pyrazol-1-yl)-2-methylpropanenitrile

  • MDL number MFCD16134951
  • Cas Number : 1250956-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90305
Availability details for
Quantity
  • Availability
  • Add to Cart