ENA341294745 Enamine

2-[(3-fluorophenyl)amino]ethan-1-ol

  • MDL number MFCD11152954
  • Cas Number : 404-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-90310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90310
Availability details for
Quantity
  • Availability
  • Add to Cart