ENA341294766 Enamine

2-cyclohexyl-N-[2-(2,3-dihydro-1H-indol-3-yl)ethyl]acetamide

  • MDL number MFCD13621569
  • Cas Number : 1042654-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90359
Availability details for
Quantity
  • Availability
  • Add to Cart