ENA341294767 Enamine

2-(chloromethyl)-5-(2-methoxyphenyl)-1,3,4-oxadiazole

  • MDL number MFCD08753715
  • Cas Number : 754214-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86...88
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90360

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90360
Availability details for
Quantity
  • Availability
  • Add to Cart