ENA341294768 Enamine

trimethyl({2-methylimidazo[1,2-a]pyridin-3-yl}methyl)azanium iodide

  • MDL number MFCD20441651
  • Cas Number : 6188-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90361
Availability details for
Quantity
  • Availability
  • Add to Cart