ENA341294774 Enamine

4-(2,3-dimethylphenyl)piperidin-4-ol hydrochloride

  • MDL number MFCD20441653
  • Cas Number : 1354960-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260...262
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90369

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90369
Availability details for
Quantity
  • Availability
  • Add to Cart