ENA341294776 Enamine

2-amino-2-(oxan-4-yl)acetonitrile hydrochloride

  • MDL number MFCD20441654
  • Cas Number : 1354963-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90371
Availability details for
Quantity
  • Availability
  • Add to Cart