ENA341294780 Enamine

3-(cyclopropylmethoxy)piperidine

  • MDL number MFCD08687523
  • Cas Number : 946725-59-3
  • Molecular Weight : 155.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90375
Availability details for
Quantity
  • Availability
  • Add to Cart