ENA341294783 Enamine

1-(2-fluorophenyl)-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD12192695
  • Cas Number : 1153042-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-90385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90385
Availability details for
Quantity
  • Availability
  • Add to Cart