ENA341294787 Enamine

2-(piperidin-4-ylsulfanyl)pyridine dihydrochloride

  • MDL number MFCD03840145
  • Cas Number : 99202-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90390
Availability details for
Quantity
  • Availability
  • Add to Cart