ENA341294793 Enamine

2-chloro-5-(isocyanatomethyl)thiophene

  • MDL number MFCD20295436
  • Cas Number : 1108713-12-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90397
Availability details for
Quantity
  • Availability
  • Add to Cart