ENA341294795 Enamine

3-(2,5,6-trimethyl-3-oxo-2,3-dihydropyridazine-4-carbonyl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD13555542
  • Cas Number : 1218015-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90400
Availability details for
Quantity
  • Availability
  • Add to Cart