ENA341294796 Enamine

2-methanesulfinyl-1-phenylethan-1-amine

  • MDL number MFCD20381897
  • Cas Number : 1343837-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90401

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90401
Availability details for
Quantity
  • Availability
  • Add to Cart