ENA341294808 Enamine

2-(morpholin-3-yl)-1-phenylethan-1-one hydrochloride

  • MDL number MFCD20441659
  • Cas Number : 1354960-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90475
Availability details for
Quantity
  • Availability
  • Add to Cart