ENA341294811 Enamine

morpholine-3-carbonitrile

  • MDL number MFCD09863124
  • Cas Number : 97039-63-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76...78
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90478

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-90478
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart