ENA341294812 Enamine

[4-(4-methoxyphenyl)phenyl]methanamine hydrochloride

  • MDL number MFCD08544236
  • Cas Number : 82487-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 298...300
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90479

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90479
Availability details for
Quantity
  • Availability
  • Add to Cart