ENA341294818 Enamine

(1H-1,3-benzodiazol-5-yl)(phenyl)methanamine dihydrochloride

  • MDL number MFCD20441664
  • Cas Number : 1354951-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-90486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90486
Availability details for
Quantity
  • Availability
  • Add to Cart