ENA341294819 Enamine

1-(piperidin-4-yl)-1,3-diazinan-2-one hydrochloride

  • MDL number MFCD20441665
  • Cas Number : 1354954-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90487
Availability details for
Quantity
  • Availability
  • Add to Cart