ENA341294822 Enamine

1-[1-(2,4-difluorophenyl)propyl]piperazine

  • MDL number MFCD05187858
  • Cas Number : 518971-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90501

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90501
Availability details for
Quantity
  • Availability
  • Add to Cart