ENA341294829 Enamine

4-(1-methyl-1H-pyrrol-2-yl)butan-2-amine

  • MDL number MFCD20441667
  • Cas Number : 1354960-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-90510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90510
Availability details for
Quantity
  • Availability
  • Add to Cart