ENA341294847 Enamine

2-(2,5-dimethylphenyl)-5-methylmorpholine hydrochloride

  • MDL number MFCD20441677
  • Cas Number : 1354960-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265...267
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90532

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90532
Availability details for
Quantity
  • Availability
  • Add to Cart