ENA341294889 Enamine

morpholine-3-carboxylic acid hydrochloride

  • MDL number MFCD01318649
  • Cas Number : 66937-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 239...241
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-90597
Availability details for
Quantity
  • Availability
  • Add to Cart