ENA341294894 Enamine

2-methyl-3-(methylamino)propan-1-ol

  • MDL number MFCD13249132
  • Cas Number : 42142-56-3
  • Molecular Weight : 103.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90603
Availability details for
Quantity
  • Availability
  • Add to Cart