ENA341294895 Enamine

2,5-dimethylmorpholine-4-sulfonyl chloride

  • MDL number MFCD12134803
  • Cas Number : 1216297-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90604
Availability details for
Quantity
  • Availability
  • Add to Cart