ENA341294898 Enamine

2,8-diazaspiro[5.5]undecane

  • MDL number MFCD11043254
  • Cas Number : 180-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-90607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90607
Availability details for
Quantity
  • Availability
  • Add to Cart