ENA341294899 Enamine

1-{[3-(2,2,2-trifluoroethyl)-1,2,4-oxadiazol-5-yl]methyl}-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD20441704
  • Cas Number : 1354957-62-2
  • Molecular Weight : 283.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-{[3-(2,2,2-trifluoroethyl)-1,2,4-oxadiazol-5-yl]methyl}-1H-pyrazol-4-ylamine hydrochloride
Inchi Code 1S/C8H8F3N5O.ClH/c9-8(10,11)1-6-14-7(17-15-6)4-16-3-5(12)2-13-16;/h2-3H,1,4,12H2;1H
Inchi Key BQSVIYIBQJWCEZ-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-90608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90608
Availability details for
Quantity
  • Availability
  • Add to Cart